Anti-PSMB9 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
PW8840-0025 PW8840-0100
89162-238EA 260.7 CAD
89162-238 89162-240
Anti-PSMB9 Mouse Monoclonal Antibody
Antibodies
The proteasome is widely recognised as the central enzyme of non-lysosomal protein degradation. It is responsible for intracellular protein turnover and it is also critically involved in many regulatory processes and, in higher eukaryotes, in antigen processing. The 26S proteasome is the key enzyme of the ubiquitin/ATP-dependent pathway of protein degradation. The catalytic core of this unusually large (2000kDa, 450Å in length) complex is formed by the 20S proteasome, a barrel shaped structure shown by electron microscopy to comprise of four rings each containing seven subunits. Based onMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol PSMB9 PSMB9
Antigen name Proteasome 20S Beta 9 Proteasome 20S Beta 9
Environmentally Preferable
Antigen synonyms macropain) subunit|proteasome-related gene 2|low molecular mass protein 2|9 (large multifunctional peptidase 2)|beta type|macropain chain 7|proteasome catalytic subunit 1i|proteasome beta 9 subunit|proteasome (prosome|RING12|MGC70470|LMP2|proteasome chain 7|beta1i|PSMB6i|proteasome subunit beta 6i|multicatalytic endopeptidase complex chain 7|PSMB9 macropain) subunit|proteasome-related gene 2|low molecular mass protein 2|9 (large multifunctional peptidase 2)|beta type|macropain chain 7|proteasome catalytic subunit 1i|proteasome beta 9 subunit|proteasome (prosome|RING12|MGC70470|LMP2|proteasome chain 7|beta1i|PSMB6i|proteasome subunit beta 6i|multicatalytic endopeptidase complex chain 7|PSMB9
Antibody type Primary Primary
Clonality Monoclonal Monoclonal
Reactivity Human Human
Host Mouse Mouse
Isotype IgG1 IgG1
Western blot Yes Yes
ImmunoChemistry Yes Yes
Form Liquid. Ascites containing 10mM sodium azide.
Purification Partially purified

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR