Anti-Proteasome 19S Rpt3/S6B Subunit Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
PW8765-0025 PW8765-0100
89162-200EA 300.2 CAD
89162-200 89162-202
Anti-Proteasome 19S Rpt3/S6B Subunit Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human and rabbit proteasome 26S (19S regulator complex) (Rpt3/S6b subunit).

Type: Primary
Antigen: PROTEASOME 19S RPT3/S6B SUBUNIT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR