Anti-Proteasome 19S Rpt1/S7 Subunit Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW8255-0025 PW8255-0100
89162-100EA 222.78 CAD
89162-100 89162-102
Anti-Proteasome 19S Rpt1/S7 Subunit Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Western Blot

Recognizes yeast proteasome 26S (19S regulator complex) (Rpt1/S7 subunit).

Type: Primary
Antigen: PROTEASOME 19S RPT1/S7 SUBUNIT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR