Anti-Proteasome 19S Rpn7/S10A Subunit Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW8225-0025 PW8225-0100
89162-076EA 297.04 CAD
89162-076 89162-078
Anti-Proteasome 19S Rpn7/S10A Subunit Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunoprecipitation, Western Blot

Recognizes human and yeast proteasome 26S (19S regulator complex) (Rpn7/S10a subunit).

Type: Primary
Antigen: PROTEASOME 19S RPN7/S10A SUBUNIT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR