Anti-Ubiquitin Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Enzo Life Sciences
PW0605-0025 PW0605-0100
89161-926EA 338.12 CAD
89161-926 89161-928
Anti-Ubiquitin Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Modification of proteins by addition of K63-linked polyubiquitin chains is implicated in a variety of cellular events, including DNA repair, signal transduction and receptor endocytosis.

  • For Western Blot

Recognizes K63-linked polyubiquitin in wide range of species. Does not cross-react with any other isopeptide-linked (K6, K11, K27, K29, K33, or K48) polyubiquitinylated species or free ubiquitin.

Type: Primary
Antigen: UBIQUITIN
Clonality: Monoclonal
Clone:
Conjugation: HRP
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Species Independent
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR