Cantharidic acid ≥98% (by TLC)
Supplier: Enzo Life Sciences
BML-PR106-0010
BML-PR106-0050
89161-762EA
135.88
CAD
89161-762
89161-764
Cantharidic acid ≥98% (by TLC)
Cantharidic acid
PP2A inhibitor
Potent and selective inhibitor of PP2A, IC50=53nM (IC50 for PP1=562nM). Useful for inhibiting PP1 and PP2A during purification of phosphorylated proteins.
Formula:
C₁₀H₁₄O₅ |
MDL Number:
MFCD00047137 CAS Number: 28874-45-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...