δ-methrin ≥99%

Supplier: Enzo Life Sciences

BML-PR101-0010 BML-PR101-0050
89161-748EA 140.62 CAD
89161-748 89161-750
δ-methrin ≥99%
Deltamethrin

Calcineurin inhibitor


Synthetic type II pyrethroid. Potent calcineurin (protein phosphatase 2B) inhibitor IC50~0.03 nM. Causes an increase in neurotransmitter release at synapses and also increases intrasynaptosomal Ca2+ levels. Serious doubts have been raised about the efficacy of this inhibitor. May also have varied effects on ion channels.

Formula: C₂₂H₁₉Br₂NO₃
MDL Number: MFCD00870122
CAS Number: 52918-63-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be398de600492ac57e01fb6379dbca5cc312f802&vid=3294ee5b7eb59938cd80b7f79b918c55b92546b3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR