Sivelestat sodium ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-PI157-0010 BML-PI157-0050
89161-706EA 203.82 CAD
89161-706 89161-708
Sivelestat sodium ≥98% (by HPLC)
Sivelestat sodium

Potent and selective leukocyte elastase inhibitor (IC50=44nM). Displays various anti-inflammatory activities including attenuation of LTB4-induced neutrophil migration and serum cytokine levels. Reduces acute inflammatory lung injury in vivo.


  • Elastase inhibitor


White solid.


Soluble in DMSO (>25mg/ml), ethanol (20mg/ml) or water (2mg/ml, warm). 

Formula: C₂₀H₂₁N₂NaO₇S·4H₂O
Storage Temperature: Ambient
MDL Number: MFCD01937969
CAS Number: 201677-61-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c788ae6dce47c213174188d560e27c0325e58da9&vid=911cf85e73f486e6ea5ec2c770d77c4e7d48948a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR