PKSI-527 ≥95% (by HPLC)

Supplier: Enzo Life Sciences

BML-PI147-0005 BML-PI147-0025
89161-692EA 237 CAD
89161-692 89161-694
PKSI-527 ≥95% (by HPLC)
PKSI-527

Highly selective plasma kallikrein inhibitor (Ki = 0.81 µM). Can be used for affinity purification of kalikrein. Suppresses collagen-induced arthritis in a mouse model. Synergizes with NO donors producing antithrombotic effects ex vivo.


  • Kallikrein inhibitor


White solid.


Soluble in water (5 mg/ml).

Formula: C₂₅H₃₁N₃O₄·HCl
Storage Temperature: Freezer
MDL Number: MFCD01745772
CAS Number: 128837-71-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=962f3bd8d4dfc29235977c80d9da4809873a8adf&vid=3b2d0a093e5399d5e982f156797ac49a19407174&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR