Gabexate mesylate ≥98% (by HPLC)
Supplier: Enzo Life Sciences
A competitive, reversible inhibitor of serine proteases tryptase (Ki=3.4nM), thrombin (Ki=500nM), uPA (Ki=1.3µM), plasmin (Ki=1.5µM), trypsin (Ki=2µM), factor Xa (Ki=2µM). At higher concentrations it inhibits kallikrein-1 (Ki=2.6mM), C1 esterase (IC50=520µM), as well as phospholipase A2 (IC50=500µM), rat iNOS (Ki=5mM) and cNOS (Ki=150µM), and swine kidney copper amine oxidase (Ki=27µM), but not chymotrypsin. Has been used in tissue culture and in vivo. In clinical use for pancreatitis.
- Protease inhibitor
White solid.
Soluble in water (25 mg/ml).
Formula:
C₁₆H₂₃N₃O₄·CH₃SO₃H Storage Temperature: Freezer |
MDL Number:
MFCD00210299 CAS Number: 56974-61-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...