Nafamostat mesylate ≥98%
Supplier: Enzo Life Sciences
Synonyms:
6-Amidino-2-naphthyl 4-Guanidinobenzoate Bis(methanesulfonate), 4-Guanidinobenzoic acid 6-Amidino-2-naphthyl Ester Bis(methanesulfonate), 4-Guanidinobenzoic acid 6-Amidino-2-naphthyl Ester Bis(methanesulphonate), 6-Carbamimidoylnaphthalen-2-yl 4-guanidinobenzoate dimethanesulfonate, 6-Carbamimidoylnaphthalen-2-yl 4-guanidinobenzoate dimethanesulphonate, 6-Amidino-2-naphthyl 4-Guanidinobenzoate Bis(methanesulphonate)
Tryptase inhibitor
Competitive, reversible broad-spectrum serine proteinase inhibitor that can be a used as a specific tryptase inhibitor at very low concentrations. Has been used in vitro and in vivo.
Formula:
C₁₉H₁₇N₅O₂ MW: 539.58 g/mol |
MDL Number:
MFCD00941430 CAS Number: 82956-11-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...