P32/98 ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-PI142-0010 BML-PI142-0050
89161-674EA 248.06 CAD
89161-674 89161-676
P32/98 ≥98% (by TLC)
P32/98

DPPIV/DPP4 inhibitor


P32/98 (Ile-thiazolidide) is a specific, competitive transition-state substrate analog inhibitor of dipeptidyl peptidase IV (DPIV; DPPIV; CD26), with a Ki of 130 nM. Useful in diabetes research. Has been used in vivo and in tissue culture.

Formula: C₁₃H₂₂N₂O₅S
MDL Number: MFCD22741399
CAS Number: 136259-20-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb3c9311a666c694d9eadc9c6c6b84abacc347da&vid=96c3bb890b7bf9fbda9232efafbf57bf452bea75&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR