BML-244 ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-PI140-0005 BML-PI140-0025
89161-670EA 153.26 CAD
89161-670 89161-672
BML-244 ≥98% (by TLC)
BML-244

A highly potent, cell-permeable inhibitor of cathepsin K (IC50=51 nM).


  • Cathepsin inhibitor


Oil.


Soluble in DMSO (10mg/ml) or 100% ethanol (12mg/ml).

Formula: C₁₁H₂₁NO₃
Storage Temperature: Freezer
CAS Number: 104062-70-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aded6fe26cd86829e9bd6355e91a3ae579c31112&vid=01aa23f7f97e737283f16ccd3ab81002d7f8f13f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR