Z-Phe-Ala-Fmk
Supplier: Enzo Life Sciences
BML-PI138-0001
BML-PI138-0005
89161-664EA
254.38
CAD
89161-664
89161-666
Z-Phe-Ala-Fmk
Proteins and Peptides
Caspase inhibitor
Irreversible inhibitor of cathepsins B, L, and S, and likely cathepsin H, as well as cruzain and papain. Has been shown to inhibit effector, but not initator, caspases in vitro, and suppress some forms of apoptosis. Has been used in vivo.