MDL-28170

Supplier: Enzo Life Sciences

BML-PI130-0010 BML-PI130-0050
89161-644EA 135.88 CAD
89161-644 89161-646
MDL-28170
Proteins and Peptides

A potent and selective cell permeable calpain inhibitor. Inhibits oxidative damage-induced apoptosis in PC12 cells and capsaicin-induced apoptosis in dorsal root ganglion neurons. It protects against anoxia in rat optic nerves and reduces neuronal damage in an animal model of global cerebral ischemia(50 mg/kg). In isolated hippocampal pyramidal neurons, it inhibits Ca²⁺-induced suppression of neurite outgrowth (complete inhibition at 20 µM). Calpain I Ki = 8 nM; Cathepsin B Ki = 24 nM in isolated enzyme assays. IC50 = 0.3 µM in intact cell assay.


  • Calpain and cathepsin inhibitor


Soluble in DMSO (200 mM).

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR