Boc-Gly-Val-Val-CHO
Supplier: Enzo Life Sciences
BML-PI128-0005
BML-PI128-0025
89161-636EA
244.9
CAD
89161-636
89161-638
Boc-Gly-Val-Val-CHO
Proteins and Peptides
gamma-secretase inhibitor
A novel substrate-based (gamma-40 site) aldehyde inhibitor of gamma-secretase. In whole cells overexpressing amloid ß protein precursor, Boc-GVV-CHO dose-dependently inhibited amyloid ß cleavage. gamma-secretase is an important therapeutic target for strategies designed to lower amyloid ß production.