PPACK
Supplier: Enzo Life Sciences
BML-PI117-0005 BML-PI117-0025
89161-612EA 314.42 CAD
89161-612 89161-614
PPACK
Proteins and Peptides
Potent irreversible thrombin inhibitor. Inhibits thrombin activation of progelatinase A. Pretreatment with PPACK inhibits thrombin-induced cerebral thromboembolism in vivo.
  • Thrombin inhibitor
White to off-white solid.
Soluble in water.