NNGH ≥99% (by TLC)

Supplier: Enzo Life Sciences

BML-PI115-0005 BML-PI115-0025
89161-604EA 140.62 CAD
89161-604 89161-606
NNGH ≥99% (by TLC)
NNGH

MMP inhibitor


Potent inhibitor of human stromelysin-1 (MMP-3), Ki=0.13 µM. Also inhibits MMP-1, MMP-2, MMP-7, MMP-8, MMP-9, MMP-10, MMP-12 (metalloelastase), MMP-13, MMP-14, and MMP-20.

MDL Number: MFCD02179370
CAS Number: 161314-17-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20648b8423158e8108208ab869f765e51d97a102&vid=62c3b6e86adf154f0db8e8ef3b95fcc192b09fa3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR