Prolyl endopeptidase inhibitor ≥97% (by HPLC)
Supplier: Enzo Life Sciences
BML-PI112-0005
BML-PI112-0025
89161-596EA
151.68
CAD
89161-596
89161-598
Prolyl endopeptidase inhibitor ≥97% (by HPLC)
Prolyl endopeptidase inhibitor
A potent and selective inhibitor of prolyl endopeptidase (PEP/PREP/POP, EC 3.4.21.26). Ki=0.35 nM for mouse brain enzyme. Inhibits the cleavage of arginine-vasopressin, gonadotropin-releasing hormone, and other peptides.
- Endopeptidase inhibitor
White solid.
Soluble in DMSO.
Formula:
C₁₈H₂₂N₂O₄ Storage Temperature: Freezer |
MDL Number:
MFCD00890691 CAS Number: 86925-97-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...