Phosphoramidon
Supplier: Enzo Life Sciences
BML-PI111-0005
BML-PI111-0025
89161-592EA
336.54
CAD
89161-592
89161-594
Phosphoramidon
Proteins and Peptides
Neprilysin inhibitor
Inhibits neutral endopeptidase 24.11 (NEP, neprilysin; EC 3.4.24.11) (Ki=2nM), a cell surface zinc metalloprotease which cleaves bioactive peptides such as bradykinin, neurotensin, and enkephalin. Also inhibits neprilysin 2 (Ki~2nM). Potent inhibitor of endothelin-converting enzyme (ECE) (IC50=0.68μM).