E-64-D
Supplier: Enzo Life Sciences
BML-PI107-0001
BML-PI107-0005
89161-582EA
146.94
CAD
89161-582
89161-584
E-64-D
Proteins and Peptides
Calpain and cathepsin inhibitor
Cell-permeable, ethyl ester of E-64-c. The ester is hydrolyzed by intracellular esterases to produce E-64-c. Inhibits calpain and cathepsins B, H, and L. Inhibits degradation of autophagic cargo inside autophagolysosomes.