Lactacystin ≥99%
Supplier: Enzo Life Sciences
Key inhibitor for use in proteasome research.
A potent and selective irreversible proteasome inhibitor. Specifically inhibits 20S proteasome (MCP; multicatalytic complex). Blocks proteasome activity by targeting the catalytic beta-subunit. Has no effect on serine or cysteine proteases. Induces apoptosis inhuman monoblast U937 cells. Inhibits NF-kappaB activation by inhibiting IkappaB degradation (IC50=10 µM). Inhibits the ubiquitin proteasome pathway in cell culture. Inhibits cathepsin A. Upregulates HSP70 and HSP22, suggesting an initial neuroprotective pathway. Induces neurite outgrowth in Neuro 2A mouse neuroblastoma cells.
Synthetic
Formula:
C₁₅H₂₄N₂O₇S |
CAS Number:
133343-34-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...