13,14-Dihydro-prostaglandin E1 ≥98% (by TLC)
Supplier: Enzo Life Sciences
Synonyms:
13,14-Dihydro-PGE1
BML-PG048-0001
BML-PG048-0010
89161-522EA
213.3
CAD
89161-522
89161-524
13,14-Dihydro-prostaglandin E1 ≥98% (by TLC)
13,14-Dihydro-prostaglandin E1
Circulating metabolite of PGE1 in man. Vasodilator equipotent with PGE12. Inhibits human platelet aggregation. Antimitotic activity.
- Prostanoid receptor ligand
Colorless to pale yellow oil
Formula:
C₂₀H₃₆O₅ MW: 356.5 g/mol Storage Temperature: Ultra-Low Temperature Freezer (–80°C) |
CAS Number:
19313-28-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...