U-46619 ≥98%
Supplier: Enzo Life Sciences
|
Warning
|
BML-PG023-0001
BML-PG023-0010
89161-478EA
282.82
CAD
89161-478
89161-480
U-46619 ≥98%
U-46619
Prostaglandin H₂/thromboxane A₂ receptor agonist; stable analog of thromboxane A₂. Induces platelet aggregation at 32 nM and activates p38 MAPK.
Soluble in 100% ethanol, DMSO, dimethyl formamide (100 mg/ml) or PBS, pH 7.2 (2 mg/ml).
Synonyms: 9,11-Dideoxy-9α,11α-methanoepoxy prostaglandin F2α, 9,11-Dideoxy-9α,11α-methanoepoxy prosta5Z,13E-dien-1-oic acid
Formula:
C₂₁H₃₄O₄ Storage Temperature: Freezer |
CAS Number:
56985-40-1 |
|
Specification Test Results
Purity | ≥98% (HPLC-ELSD) |
Formulation | Pale yellow oil |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...