Vinpocetine ≥98%

Supplier: Enzo Life Sciences

Synonyms: (41S,13aS)-Ethyl 13a-ethyl-2,3,41,5,6,13a-hexahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate

BML-PD185-0020 BML-PD185-0100
89161-372EA 118.5 CAD
89161-372 89161-374
Vinpocetine ≥98%
Vinpocetine

PDE inhibitor


Selective inhibitor of Ca2+/calmodulin-dependent phosphodiesterase (PDE I, IC50=20 µM).

Formula: C₂₂H₂₆N₂O₂
MW: 350.46 g/mol
MDL Number: MFCD00211233
CAS Number: 42971-09-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0454b3f17ca24c6a2d61d8d4245fb4d8127e4cc&vid=1151f8daf57c76ed84137d1e30eb2ef892c7d20a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR