Rolipram ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-PD175-0010 BML-PD175-0050
89161-358EA 140.62 CAD
89161-358 89161-360
Rolipram ≥98% (by TLC)
Rolipram

PDE inhibitor


Selective, cell permeable inhibitor of cAMP-specific phosphodiesterase (PDE IV, IC50=1 µM). A rolipram insensitive PDE IV subtype has been described.

Formula: C₁₆H₂₁NO₃
MDL Number: MFCD00270906
CAS Number: 61413-54-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=177c24a1c668382ffd0e475a8740725107f7bb82&vid=a98b4b08094c50ed6c52ab3efe656ed449d5c6af&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR