MBCQ ≥98%
Supplier: Enzo Life Sciences
BML-PD149-0005
BML-PD149-0025
89161-346EA
162.74
CAD
89161-346
89161-348
MBCQ ≥98%
MBCQ
Selective inhibitor of cGMP-specific phosphodiesterase (PDE V, IC50=19 nM).
- PDE inhibitor
White solid.
Soluble in DMSO.
Formula:
C₁₆H₁₂CIN₃O₂ Storage Temperature: Ambient |
CAS Number:
150450-53-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...