EHNA ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-PD129-0010 BML-PD129-0050
89161-330EA 127.98 CAD
89161-330 89161-332
EHNA ≥98% (by HPLC)
EHNA

Phosphodiesterase PDE II inhibitor, IC50=0.8 µM. Also inhibits adenosine deaminase, Ki=4 nM.


  • PDE inhibitor


White solid.


Soluble in water (25mg/ml).

Formula: C₁₄H₂₃N₅O·HCl
Storage Temperature: Ambient
CAS Number: 51350-19-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58a71c6af5c7fe67d0d2afcc08f30b0c4e1f701e&vid=e8fc146503bcaa0828b3cf7afed1fbcc327b6db1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR