Cilostazol ≥98%

Supplier: Enzo Life Sciences

Synonyms: 6-(4-(1-Cyclohexyl-1H-tetrazol-5-yl)butoxy)-3,4-dihydroquinolin-2(1H)-one, 6-[4-(1-Cyclohexyl-1H-tetrazol-5-yl)butoxy]-3,4-dihydro-2(1H)-quinolinone

BML-PD127-0010 BML-PD127-0050
89161-326EA 143.78 CAD
89161-326 89161-328
Cilostazol ≥98%
Cilostazol

PDE inhibitor


Cilostazol is a PDE3A inhibitor (0.2µM) and a unique antithrombotic agent. It inhibits cell death by activating maxi-K channels.

Formula: C₂₀H₂₇N₅O₂
MW: 369.47 g/mol
MDL Number: MFCD00866780
CAS Number: 73963-72-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fdce6b34ed4d9e0fda9bd2620f098b479d22b0fd&vid=4f267dd4785b1b9f89327078aa49f38fbbfa62f5&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR