Cilostamide ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: N-Cyclohexyl-N-methyl-4-((2-oxo-1,2-dihydroquinolin-6-yl)oxy)butanamide

BML-PD125-0005 BML-PD125-0025
89161-322EA 140.62 CAD
89161-322 89161-324
Cilostamide ≥98% (by TLC)
Cilostamide

PDE inhibitor


Phosphodiesterase PDE3 inhibitor (IC50=5 nM). Blocks PKB/Akt signaling pathway by inhibition of Akt-activated PDE3B. Reverses the established effects of leptin on food intake and body weight. Blocks the leptin-induced tyrosine phosphorylation of STAT3 at the hypothalamic level.

Formula: C₂₀H₂₆N₂O₃
MW: 342.43 g/mol
MDL Number: MFCD00673958
CAS Number: 68550-75-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=405ee75093644e6855522068508f031e7160d498&vid=d094718acd5da88a944026b581b4297967f93778&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR