N,N,2-Trimethyl-5-nitrobenzenesulfonamide (synthetic) ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-PD120-0010 BML-PD120-0050
89161-318EA 158 CAD
89161-318 89161-320
N,N,2-Trimethyl-5-nitrobenzenesulfonamide (synthetic) ≥98% (by TLC)
N,N,2-Trimethyl-5-nitrobenzenesulfonamide

The first selective PDE7 inhibitor (Ki=180 nM).


  • PDE inhibitor


White to off-white solid.

Formula: C₉H₁₂N₂O₄S
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD03039912
CAS Number: 433695-36-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c32213fb0da3d4eeaad80fff3ea4e0e3e7dfcf7&vid=c4e4f9a2c8bb0042d082547e51d91413301d0211&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR