Bupropion hydrochloride ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-NS111-0050 BML-NS111-0250
89160-844EA 143.78 CAD
89160-844 89160-846
Bupropion hydrochloride ≥98% (by TLC)
Bupropion hydrochloride

Inhibitor of dopamine and norepinephrine transporters


Inhibits dopamine and norepinephrine transporters (Ki=2.8 and 1.4µM respectively). Inhibits the serotonin transporter at higher concentration (Ki=45µM). In clinical use as an antidepressant and for smoking cessation. Also acts as a nicotinic receptor antagonist.

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD00055209
CAS Number: 34911-55-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7dd1435947ebe26d921e463614f804fbbb39b56&vid=2a6f147884768dfaa38abbb982c8c0b371bfb341&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR