Dihydroergocristine methanesulfonate ≥98%, white solid

Supplier: Enzo Life Sciences
Danger

Synonyms: Dihydroergocristine mesylate

BML-NS108-0200 BML-NS108-1000
89160-836EA 105.86 CAD
89160-836 89160-838
Dihydroergocristine methanesulfonate ≥98%, white solid
Dihydroergocristine methanesulfonate

Serotonin receptor ligand.


  • Formulation: White solid.


Serotonin antagonist. Dopaminergic and adrenergic partial agonist. Cognition enhancer.


Soluble in water (10 mg/ml), methanol (22 mg/ml), or DMSO (25 mM).

Formula: C₃₆H₄₅N₅O₈S
Storage Temperature: Ambient
MDL Number: MFCD00153792
CAS Number: 24730-10-7
UN: 1544
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e18d62d6873c1c6a4ef309fd17951188e0bca2b3&vid=e09e82480e3f2114be69435075e522e25a426947&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR