Clozapine-N-oxide ≥99% (by HPLC), yellow solid

Supplier: Enzo Life Sciences
Danger

BML-NS105-0005 BML-NS105-0025
89160-824EA 183.28 CAD
89160-824 89160-826
Clozapine-N-oxide ≥99% (by HPLC), yellow solid
Clozapine-N-oxide

Major metabolite of clozapine. Possesses little or no activity at 5-HT receptors.


  • High Purity
  • Active in vitro and in vivo


Contains 1 equivalent of methanol.


Soluble in water (5 mg/ml).

Formula: C₁₈H₁₉ClN₄O
Storage Temperature: Ambient
MDL Number: MFCD00210190
CAS Number: 34233-69-7
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e367c1ac802e0577184f65a9ef49fa203f5a0d22&vid=2a9a06e1acb6db4974ae2b9c14eb4451604921a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥99% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR