Astemizole ≥99% (by TLC), off-white powder

Supplier: Enzo Life Sciences
Warning

Synonyms: 1-(4-Fluorobenzyl)-2-(1-[4-methoxyphenethyl]piperidin-4-yl)aminobenzimidazole, astemizole

BML-NH101-0010 BML-NH101-0050
89160-732EA 150.1 CAD
89160-732 89160-734
Astemizole ≥99% (by TLC), off-white powder
Astemizole

Histamine receptor ligand


  • Formulation: Off-white powder.


Selective histamine H1 receptor antagonist.


Soluble in chloroform (clear faint yellow solution at 100 mg/ml).

Formula: C₂₈H₃₁FN₄O
Storage Temperature: Ambient
MDL Number: MFCD00153919
CAS Number: 68844-77-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23a9f7c589b6c3e44e5540dab642079288757003&vid=089bc4efa3c57dd52ef0709087f46e55b82451a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥99% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR