Lorcainide hydrochloride ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-NA143-0010 BML-NA143-0050
89160-704EA 170.64 CAD
89160-704 89160-706
Lorcainide hydrochloride ≥98% (by TLC)
Lorcainide hydrochloride

Na+ channel blocker


Possesses both local anesthetic and antiarrhythmic properties (class 1C agent). Elevates the ventricular fibrillation threshold and protects against ouabain-induced arrhythmias in animal models. Its mechamism of action is primarly inhibition of sodium channels in a voltage- and frequency-dependent fashion. Also inhibits the microsomal Na+,K+-ATPase in a concentration-dependent manner (IC50=33.5µM) . Reduces neuronal death in cellular models of hypoxic injury when combined with a glutamate receptor antagonist and blocks aconitine-induced mortality in a rat model (ED50=30mg/kg).

Formula: C₂₂H₂₈Cl₂N₂O
MDL Number: MFCD00055196
CAS Number: 58934-46-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56293d9ffd727c46d272e5132dcad5dd436b13e3&vid=3d628f9f97572e051c14fca231e6ee3cf5fd18c0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR