Kavain (+/-) ≥98%

Supplier: Enzo Life Sciences

Synonyms: (±)-4-Methoxy-6-styryl-5,6-dihydro-α-pyrone, (+)-Kavain, (R,E)-4-Methoxy-6-styryl-5,6-dihydro-2H-pyran-2-one

BML-NA136-0050 BML-NA136-0250
89160-680EA 143.78 CAD
89160-680 89160-682
Kavain (+/-) ≥98%
Kavain (+/-)

Kavain is one constituent of kava-kava, an herbal drug of the South Pacific islands, which displays anti-convulsive, anxiolytic, analgesic and muscle relaxing properties. Kavain inhibits voltage-dependent Na+ channels in rat CA1 hippocampal neurons. It displays protective effects in the MPTP mouse model of Parkinson's disease but potentiates NMDA toxicity in the developing hippocampus.


  • Na+ channel blocker


Kavain is one constituent of kava-kava, an herbal drug of the South Pacific islands, which displays anti-convulsive, anxiolytic, analgesic and muscle relaxing properties. Kavain inhibits voltage-dependent Na+ channels in rat CA1 hippocampal neurons. It displays protective effects in the MPTP mouse model of Parkinson's disease but potentiates NMDA toxicity in the developing hippocampus.

Formula: C₁₄H₁₄O₃
MW: 230.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00467093
CAS Number: 500-64-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ff76ab43117e2a982dd60be0e9ec5c7e933fb7c&vid=47cbe47e20cc83c49b3da7698fd27f4dc5288f00&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR