Gambogic amide ≥98% (by TLC)

Supplier: Enzo Life Sciences
Danger

BML-N159-0001 BML-N159-0005
89160-604EA 237 CAD
89160-604 89160-606
Gambogic amide ≥98% (by TLC)
Gambogic amide

TrkA ligand


  • Formulation: Yellow-orange solid.


A novel small molecule NGF mimic discovered using a cell-based chemical genetic screen for TrkA agonists. Gambogic amide selectively binds to TrkA, triggers its tyrosine phosphorylation, elicits PI3-kinase/Akt and MAPK activation, provokes neurite outgrowth in PC12 cells and prevents neuronal cell death. EC50 for antiapoptotic activity is 5nM.


Soluble in DMSO (20 mg/ml) or ethanol (20 mg/ml).

Formula: C₃₈H₄₅NO₇
Melting Pt: 74…76 °C
Storage Temperature: Ambient
CAS Number: 286935-60-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ede2973f64c24278e3fee48bc4f9bcbecc0b0af7&vid=0bf45e22b3a568f469bd8d5855fb9568db7bdbd8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (TLC)
Appearance Yellow-orange solid
Melting Point 74 - 76°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR