Leukotriene A4 methyl ester ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Danger

LPA004-0050 LPA004-1000
89160-562EA 418.7 CAD
89160-562 89160-564
Leukotriene A4 methyl ester ≥98% (by HPLC)
Leukotriene A4 methyl ester

Biosynthesis in rabbit reticulocytes and human leukocytes.


  • Formulation: Liquid.


Synonym: 14,15-LTA₄-ME

Formula: C₂₁H₃₂O₃
MW: 332.5 g/mol
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
CAS Number: 73466-12-3
UN: 3082
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c65566101b374635b8647156abdadfc98d8577e6&vid=feee9760beab71c0d1f4b701844d94362288db9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)
Concentration 50 µg/ml
Formulation Oil dissolved in 2% triethylamine/hexane


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR