Mono-n-dodecyl phosphate ≥98% (by TLC), white to off-white solid
Supplier: Enzo Life Sciences
Synonyms:
Dihydrogen monolauryl phosphate
BML-LP105-0005
BML-LP105-0025
89160-550EA
188.02
CAD
89160-550
89160-552
Mono-n-dodecyl phosphate ≥98% (by TLC), white to off-white solid
Mono-n-dodecyl phosphate
Lysophosphatidic acid (LPA) receptor ligand. A specific agonist at LPA2 (EC₅₀=700 nM) and antagonist at LPA.
- Formulation: White to off-white solid.
Soluble in DMSO (15 mg/ml warm) or ethanol (2 mg/ml warm).
Formula:
C₁₂H₂₇O₄P MW: 266.318 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00013946 CAS Number: 2627-35-2 |
|
Specification Test Results
Purity | ≥98% (TLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...