(±)-Lisofylline ≥98%

Supplier: Enzo Life Sciences

Synonyms: 1-(5'-Hydroxyhexyl)-3,7-dimethylxanthine

BML-LP102-0010 BML-LP102-0050
89160-540EA 176.96 CAD
89160-540 89160-542
(±)-Lisofylline ≥98%
(±)-Lisofylline

Potent inhibitor of phosphatidic acid generation


A potent inhibitor of phosphatidic acid generation (IC50=0.6 µM). It protects mice from endotoxic shock and attenuates sepsis-induced acute lung injury in the pig. Anti-inflammatory agent. Enhances glucose-stimulated insulin secretion. Inhibits IL-12-mediated STAT4 activation.

Formula: C₁₃H₂₀N₄O₃
MW: 280.3 g/mol
MDL Number: MFCD00867573
CAS Number: 6493-06-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a02eee796393797308b9cf7aa266d67b93cc5e49&vid=21767cf94fabe7103466fd287933a84171b9ab9e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR