Lysophosphatidic acid ≥98% (by TLC)
Supplier: Enzo Life Sciences
LPA receptor ligand
- Formulation: Waxy solid.
Lysophosphatidic acid (LPA) is a multifunctional intercellular phospholipid messenger.
LPA stimulates the growth of a variety of cells including fibroblasts, vascular smooth muscle cells, endothelial cells and keratinocytes. It acts as a proliferative and anti-apoptotic factor. Ligand for LPA1 (EDG-2), LPA2 (EDG-4) and LPA3 (EDG-7) receptors. It also inhibits differentiation of neural stem cells into neurons.
Soluble in water (5 mg/ml with sonication) or DMSO (0.5 mg/ml warm).
Formula:
C₂₁H₄₀O₇P·Na Storage Temperature: Freezer |
MDL Number:
MFCD00133427 CAS Number: 325465-93-8 |
|
Specification Test Results
Purity | ≥98% (TLC) |
Appearance | Waxy solid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...