Leukotriene B4 ≥99%

Supplier: Enzo Life Sciences
Danger

Synonyms: ((5S),6Z,8E,10E,12(R),14Z)-5,12-Dihydroxyeicosatetraenoic acid, LTB4

BML-LB004-0050 BML-LB004-1000
89160-494EA 589.34 CAD
89160-494 89160-496
Leukotriene B4 ≥99%
Leukotriene B4

Prostanoid receptor ligand.


  • Formulation: Colorless liquid oil dissolved in 1ml ethanol.


Activator of PPARα. Stimulates leukocyte functions including chemokinesis, chemotaxis, lysosomal enzyme release, adhesion, stimulation of ion fluxes, superoxyde anion production and C3b receptor expression. Induces vascular permeability, NK cell activity, and broncho-constriction.

Formula: C₂₀H₃₂O₄
MW: 335.464 g/mol
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
MDL Number: MFCD00135630
CAS Number: 71160-24-2
UN: 1170
ADR: 3 II
Merck Index: 14,05455
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ccd5857b4d22057675895bd641d7c4031c8ac27&vid=dec0aaed8990cc5886bad19c5ae1aa0dae10ea48&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Concentration (50 µg size) 50 µg/ml
Concentration (1 mg size) 1 mg/ml
Formulation Oil dissolved in ethanol.
Purity ≥99% (HPLC)
Source Synthetic


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR