Leukotriene A4 methyl ester ≥95% (by HPLC)

Supplier: Enzo Life Sciences
Danger

BML-LA004-0050 BML-LA004-1000
89160-486EA 669.92 CAD
89160-486 89160-488
Leukotriene A4 methyl ester ≥95% (by HPLC)
Leukotriene A4 methyl ester

Substrate for LTA₄ hydrolase and LTC₄ synthase.


  • Formulation: Liquid.


Unstable intermediate in the biosynthesis of LTB₄ and LTC₄. The naturally occuring free acid is a substrate for LTA₄ hydrolase and LTC₄ synthase and plays a central role in transcellular leukotrieneand lipoxin biosynthesis. Mobilises Ca²⁺ in human neutrophils. Supplied as the methyl ester for greater stability.

Formula: C₂₁H₃₂O₃
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
CAS Number: 73466-12-3
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e35acead84cca3f8bdefff7ebdc0edd3f86f0661&vid=ad9e0163c14a10ac535f57b9de24e39c2b42e12e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥95% (HPLC) (remainder 11-trans isomer)
Formulation Oil dissolved in 2% triethylamine/hexane.
Concentration 1 mg/ml
Source Synthetic


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR