Bis(methylthio)gliotoxin ≥99% (by TLC)

Supplier: Enzo Life Sciences

BML-L122-0001 BML-L122-0005
89160-450EA 312.84 CAD
89160-450 89160-452
Bis(methylthio)gliotoxin ≥99% (by TLC)
Bis(methylthio)gliotoxin

Inhibits PAF-induced platelet aggregation (IC50=8.4 µM) and collagen-induced aggregation (IC50=84.2 µM) with no effect on arachidonic acid or ADP induced aggregation. Inhibits PAF-induced bronchoconstriction in vivo in the guinea-pig (69% at 1.0 mg/kg).


  • PAF receptor ligand


White solid.


Soluble in DMSO (25mg/ml) and ethanol (25mg/ml).

Formula: C₁₅H₂₀N₂O₄S₂
MW: 356.455 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00133146
CAS Number: 74149-38-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc34a3112568acfcaa30ffb606ea9ac2cc850da6&vid=4d8e7540c164a906ce8953a428a3a4f9a1a45bec&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR