Sodium dodecyl sulfate (SDS)

Supplier: Enzo Life Sciences
Danger

Synonyms: Sodium dodecyl sulfateLauryl sulfate sodium saltSodium lauryl sulfateDodecyl sulfate sodium saltDodecyl sulphuric acid sodium saltLauryl sulphate sodium saltSodium lauryl sulphateDodecyl sulphate sodium saltDodecyl sulfuric acid sodium salt

BML-KI341-0012
89160-374EA 71.1 CAD
89160-374
Sodium dodecyl sulfate (SDS)
Sodium dodecyl sulphate (SDS)

Component of the 20S proteasome drug discovery assay.

Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00036175
CAS Number: 151-21-3
UN: 2926
ADR: 4.1,II
https://ca.vwr.com/stibo/web/std.lang.all/54/91/735491.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR