Nuclear extract from HeLa cells, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-KI140-0100
89160-320EA 209 CAD
89160-320
Nuclear extract from HeLa cells, Enzo Life Sciences
Cells
Prepared by high salt extraction of HeLa nuclei (human cervical cancer cell line), this extract is a rich source of HDAC activity, useful for inhibitor screening or immunoprecipitation of HDAC containing complexes. Typically, 0.5 µl or less per well is enough for assay with the HDAC FLUOR DE LYS® fluorometric activity assay.
Formulation: Liquid in 0.1 M KCl, 20 mM HEPES/NaOH, pH 7.9, 20% (v/v) glycerol, 0.2 mM EDTA, 0.5 mM DTT, 0.5 mM PMSF.