AMC (7-Amino-4-methylcoumarin) Standard, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-KI107-0001
89160-288EA
74.26
CAD
89160-288
AMC (7-Amino-4-methylcoumarin) Standard, Enzo Life Sciences
Protein Linking and Labeling Systems
Fluorogenic chromophore. Ex(max): 380nm, Em(max): 460nm. Reference compound.