NS-1643 ≥98% (by TLC)
Supplier: Enzo Life Sciences
|
Warning
|
Synonyms:
NS1643, 1,3-Bis(2-hydroxy-5-(trifluoromethyl)phenyl)urea
BML-KC169-0010
BML-KC169-0050
89160-266EA
197.5
CAD
89160-266
89160-268
NS-1643 ≥98% (by TLC)
NS-1643
K⁺ channel activator.
- Formulation: White solid
A novel and selective HERG K⁺ channel activator (EC50=10.5 µM). Activates ERG2 channels. Displays antiarrhythmic properties in rabbit models.
Soluble in DMSO (25 mg/mL) or ethanol (25 mg/mL).
Formula:
C₁₅H₁₀F₆N₂O₃ MW: 380.2 g/mol |
MDL Number:
MFCD08705418 CAS Number: 448895-37-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity | ≥98% (TLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...