N-(1-Adamantyl)-N'-cyclohexylmorpholine-4-carboxamidine hydrochloride ≥98%

Supplier: Enzo Life Sciences

Synonyms: U-37883A

BML-KC159-0005 BML-KC159-0025
89160-238EA 176.96 CAD
89160-238 89160-240
N-(1-Adamantyl)-N'-cyclohexylmorpholine-4-carboxamidine hydrochloride ≥98%
N-(1-Adamantyl)-N'-cyclohexylmorpholine-4-carboxamidine hydrochloride

K+ channel blocker


A non-sulfonylurea inhibitor of K-ATP channels. Inhibits vascular K-ATP channels. Inhibits dopamine-modulated potassium channels.

Formula: C₂₁H₃₆ClN₃O
MW: 381.989 g/mol
MDL Number: MFCD03840615
CAS Number: 57568-80-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fd2dbe52be90266bc7ac3716eaee21d1a836524&vid=28152fad05d80669fa8419561cf02abfd67bd27f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR